Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, HEPES solution (1 mol/L)

Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 100 mL
Catalog # INVT15630056
Marketsource
Supplier:   Invitrogen
Buffer, HEPES solution (1 mol/L)
Catalog # INVT15630056
Supplier:  Invitrogen
Marketsource
CAS Number:  
Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 100 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 ml
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    HEPES buffer solution (1 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.