Vanadium(V) oxide ≥99.6%
Glass bottle for solids, 500 g
Purity: 99.6% min CAS No.: 1314-62-1 Molecular Formula: V2O5 Formula Weight: 181.88 Physical Form: -10 Mesh Density: 3.357 Melting Point: 690° Boiling Point: 1750° dec. Merck Reference: 14,9921 UN No.: 2862 MDL No.: MFCD00011457
: Vanadium pentoxide, di-Vanadium pentaoxide
- :1314-62-1
- :O₅V₂
- :MFCD00011457
- :181.88 g/mol
- :14,9921
- :1750 °C (dec.)
- :690 °C
- :3.357
- :Ambient
- :215-239-8
- :2862
- :6.1,III
- Pack type:Glass bottle for solids
- Pk:500 g