Orders LinkContactUsLinkComponent
Hose clips
9 - 10 mm, White
Catalog # BURK8678-0010
Supplier:   BURKLE
Hose clips
Catalog # BURK8678-0010
Supplier:  BURKLE
Hose clips
Hose clips
9 - 10 mm, White
Price
SFr. 5.90
Log in for availability
1 * 10 ST
Product Details & Documents
Hose clips made of Polyacetal (POM).
Specifications
  • Spanning width:
    9 - 10 mm
  • Colour:
    White
  • Seq:
    02