Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Boxes with Snap-On Lid, Multipurpose

Boxes with Snap-On Lid, Multipurpose

90 mm height, 78 mm Ø, 250 ml
Catalog # BURK6210-0250
Supplier:   BURKLE
Boxes with Snap-On Lid, Multipurpose
Catalog # BURK6210-0250
Supplier:  BURKLE
Boxes with Snap-On Lid, Multipurpose
Boxes with Snap-On Lid, Multipurpose

90 mm height, 78 mm Ø, 250 ml

Price
SFr. 5.40
Log in for availability
1 * 1 ST
Product Details & Documents

Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (−40 °C up to briefly +95 °C).
PE, translucent.

  • Can be deep frozen
  • No crevices
  • Suitable for use with foodstuffs
Specifications
  • Capacity:
    250 ml
  • Description:
    90 mm height, 78 mm Ø
  • Seq:
    01