Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Boxes with Snap-On Lid, Multipurpose

Boxes with Snap-On Lid, Multipurpose

110 mm height, 136 mm Ø, 1000 ml
Catalog # BURK6210-1001
Supplier:   BURKLE
Boxes with Snap-On Lid, Multipurpose
Catalog # BURK6210-1001
Supplier:  BURKLE
Boxes with Snap-On Lid, Multipurpose
Boxes with Snap-On Lid, Multipurpose

110 mm height, 136 mm Ø, 1000 ml

Price
SFr. 8.00
Log in for availability
1 * 1 ST
Product Details & Documents

Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (−40 °C up to briefly +95 °C).
PE, translucent.

  • Can be deep frozen
  • No crevices
  • Suitable for use with foodstuffs
Specifications
  • Capacity:
    1000 ml
  • Description:
    110 mm height, 136 mm Ø
  • Seq:
    03