Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buckets

Buckets

Bucket, with graduation and handle, White, 5 L, 240 mm
Catalog # BRND71772
Supplier:   Brand
Buckets
Catalog # BRND71772
Supplier:  Brand
Buckets
Buckets

Bucket, with graduation and handle, White, 5 L, 240 mm

Price
SFr. 22.10
Log in for availability
1 * 1 ST
Product Details & Documents

HDPE, white.

  • Good chemical and temperature resistance
  • Scale with 1 litre steps
  • Without spout and lid (Please order push-on lid seperately)

For industrial and laboratory applications. With handle.

Specifications
  • Capacity:
    5 L
  • Colour:
    White
  • Description:
    Bucket, with graduation and handle
  • Height:
    240 mm
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Push-on lid, LDPE
    Push-on lid, LDPE
    BRND71776
    10 l buckets
    Log in for availability
    1 * 1 ST - SFr. 7.30

Product Information

  • Catalog #
    For
    Availability
    Price
  • Push-on lid, LDPE
    Push-on lid, LDPE
    BRND71771
    5 L buckets
    Log in for availability
    1 * 1 ST - SFr. 6.70

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.