Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Bulb pipettes, BLAUBRAND®, two marks, class AS

1 ml, 0,008 ± ml, 300 mm, Blue
Catalog # 612-2727
Supplier:   Brand
Bulb pipettes, BLAUBRAND®, two marks, class AS
Catalog # 612-2727
Supplier:  Brand
Bulb pipettes, BLAUBRAND®, two marks, class AS
Bulb pipettes, BLAUBRAND®, two marks, class AS

1 ml, 0,008 ± ml, 300 mm, Blue

Price
SFr. 10.50
Log in for availability
1 * 1 ST
Product Details & Documents

AR-Glas®.

  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Including one batch certificate

DIN EN ISO 648

Certifications: DIN EN ISO 648; DE-M marking.

Specifications
  • Colour code:
    Blue
  • Capacity:
    1 ml
  • Length:
    300 mm
  • Seq:
    02
  • Tolerance:
    0,008 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.