Volumetric flasks, class A, blue markings
1000 ml, 0,4 ± ml
Catalog # BRND2.37053
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 3.3, with beaded rim.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Total order from one batch, batch certificate included
: DIN EN ISO 1042
Specifications
- Capacity:1000 ml
- Seq:05
- Tolerance:0,4 ± ml