Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

115 mm, 14 mm, 150 mm
Catalog # BRND148240
Supplier:   Brand
Funnels with short stem
Catalog # BRND148240
Supplier:  Brand
Funnels with short stem
Funnels with short stem

115 mm, 14 mm, 150 mm

Price
SFr. 18.90
Log in for availability
1 * 1 ST
Product Details & Documents

PP, translucent.

  • External flutings allow air to escape during filtration
  • Excellent chemical resistance, autoclavable
  • Suitable for use with standard filter papers

For rapid filtration.

Specifications
  • Funnel O-Ø:
    150 mm
  • Stem O-Ø:
    14 mm
  • Stem length:
    115 mm