Funnels with short stem
63 mm, 9 mm, 65 mm
BRND148220
: Brand
PP, translucent.
- External flutings allow air to escape during filtration
- Excellent chemical resistance, autoclavable
- Suitable for use with standard filter papers
For rapid filtration.
- Funnel O-Ø:65 mm
- Stem O-Ø:9 mm
- Stem length:63 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



