Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

44 mm, 5 mm, 45 mm
Catalog # BRND148215
Supplier:   Brand
Funnels with short stem
Catalog # BRND148215
Supplier:  Brand
Funnels with short stem
Funnels with short stem

44 mm, 5 mm, 45 mm

Price
SFr. 95.10
Log in for availability
1 * 20 ST
Product Details & Documents

PP, translucent.

  • External flutings allow air to escape during filtration
  • Excellent chemical resistance, autoclavable
  • Suitable for use with standard filter papers

For rapid filtration.

Specifications
  • Funnel O-Ø:
    45 mm
  • Stem O-Ø:
    5 mm
  • Stem length:
    44 mm
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.