Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Stirring rod, round ended, with steel core, PTFE, 6 mm, 400 mm
Catalog # 441-0216
Supplier:   Brand
Stirring rods
Catalog # 441-0216
Supplier:  Brand
Stirring rods
Stirring rods

Stirring rod, round ended, with steel core, PTFE, 6 mm, 400 mm

Price
SFr. 35.70
Log in for availability
1 * 1 ST
Product Details & Documents

Stirring rods.

Specifications
  • Ø:
    6 mm
  • Description:
    Stirring rod, round ended, with steel core
  • Length:
    400 mm
  • Material:
    PTFE
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.