Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Dressing jars, with lid

6000 ml, 200 mm, 200 mm
  BOCH8693
 :   BOCHEM
Dressing jars, with lid
  BOCH8693
 :  BOCHEM
Dressing jars, with lid
Dressing jars, with lid

6000 ml, 200 mm, 200 mm

 
SFr. 185.00
   
1 * 1 ST
 

Jar made of stainless steel, with knob lid.

  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
 
  • Ext.Ø:
    200 mm
  • Capacity:
    6000 ml
  • Height:
    200 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.