Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dressing jars, with lid

Dressing jars, with lid

1200 ml, 120 mm, 120 mm
Catalog # BOCH8587
Supplier:   BOCHEM
Dressing jars, with lid
Catalog # BOCH8587
Supplier:  BOCHEM
Dressing jars, with lid
Dressing jars, with lid

1200 ml, 120 mm, 120 mm

Price
SFr. 76.50
Log in for availability
1 * 1 ST
Product Details & Documents

Jar made of stainless steel, with knob lid.

  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Specifications
  • Ext.Ø:
    120 mm
  • Capacity:
    1200 ml
  • Height:
    120 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.