Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dressing jars, with lid

Dressing jars, with lid

700 ml, 100 mm, 100 mm
Catalog # BOCH8585
Supplier:   BOCHEM
Dressing jars, with lid
Catalog # BOCH8585
Supplier:  BOCHEM
Dressing jars, with lid
Dressing jars, with lid

700 ml, 100 mm, 100 mm

Price
SFr. 67.70
Log in for availability
1 * 1 ST
Product Details & Documents

Jar made of stainless steel, with knob lid.

  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Specifications
  • Ext.Ø:
    100 mm
  • Capacity:
    700 ml
  • Height:
    100 mm
  • Seq:
    03