Tripods
With welded feet, stainless steel, 140×210 mm
Catalog # BOCH5202
Supplier: BOCHEM
Jump to:
Product Details & Documents
Tripods made from stainless steel or tempered cast iron (zincked).
- With welded or screwed feet
Specifications
- Int.Ø×H:140×210 mm
- Description:With welded feet, stainless steel