Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pharmacist spoons

Pharmacist spoons

Spoon with hole, 200 mm, 30×26 mm
Catalog # 231-1281
Supplier:   BOCHEM
Pharmacist spoons
Catalog # 231-1281
Supplier:  BOCHEM
Pharmacist spoons
Pharmacist spoons

Spoon with hole, 200 mm, 30×26 mm

Price
SFr. 7.50
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spoons for a wide range of uses.

  • Narrow shape with extra long, flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Description:
    Spoon with hole
  • Length:
    200 mm
  • Spoon size:
    30×26 mm