Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Curved, extra sharp, 120 mm
Catalog # 232-0010
Supplier:   BOCHEM
Forceps, precision
Catalog # 232-0010
Supplier:  BOCHEM
Forceps, precision
Forceps, precision
Curved, extra sharp, 120 mm
Price
12.50 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production etc.
  • Extremely sharp tips without grooves
  • Reverse action; closed normally, squeeze to open
Specifications
  • Type:
    Curved, extra sharp
  • Length:
    120 mm