(2-Hydroxylpropyl)-β-cyclodextrine 97%
Glass bottle, 100 g
: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
- :128446-35-5
- :C₆₃H₁₁₂O₄₂
- :MFCD16621721
- :278 °C (dec.)
- :Ambient
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Water:≤7.5 % (K.F.)
- HPLC:≥96.0 % (On dry substance)
- NMR:2 to 6 (degree of substitution)
- Pack type:Glass bottle
- Pk:100 g



