(S)-(+)-2-Hydroxyisovaleric acid 99+%
Glass bottle, 1 g
Anbieter: Thermo Fisher Scientific
(S)-(+)-2-Hydroxyisovaleric acid 99+%
Katalognr. ACRO269900010
Anbieter: Thermo Fisher Scientific
CAS Nummer: 17407-55-5
Direkt zu
Product Details & Documents
Synonyme: (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid
- CAS Nummer:17407-55-5
- Formel:C₅H₁₀O₃
- MDL-Nummer:MFCD00066443
- Molekulargewicht:118.13 g/mol
- Schmelzender Platingehalt:65…68 °C
- Lagertemperatur:Refrigerator
Dokumente
ACRO26990
Sicherheitsdatenblatt (SDS)
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Melting point:65 to 70 °C
- GC:≥99.0 %
- Specific optical rotation:+15° to +19° (24 °C, 589 nm) (c=2, CHCl3)
- Titration with NaOH:≥99.0 %
Technische Daten
- Pack type:Glass bottle
- Pk:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



