ContactUsLinkComponent
Stirring bar, micro (flea), magnetic, PTFE, white, size 5x2mm, small sizes for stirring small volumes in vessels such as vials, tube s, and gradient makers. 1 * 1 items
Catalog # BELAF371190005
Stirring bar, micro (flea), magnetic, PTFE, white, size 5x2mm, small sizes for stirring small volumes in vessels such as vials, tube s, and gradient makers. 1 * 1 items
Catalog # BELAF371190005
Frequently Bought Together