Orders LinkContactUsLinkComponent
Sieves
Stainless steel sieve, 1,00 mm, Sieve MF 1.0
Katalognr. 412-1136
Anbieter:   IKA
Sieves
Katalognr. 412-1136
Anbieter:  IKA
Sieves
Sieves
Stainless steel sieve, 1,00 mm, Sieve MF 1.0
Preis
SFr. 483.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
Technische Daten
  • Type:
    Sieve MF 1.0
  • Description:
    Stainless steel sieve, 1,00 mm
  • Seq:
    08