Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Transfer capillaries

80 mm, 2,35 mm, 1,55
  HIRS9202380
 :   HIRSCHMANN
Transfer capillaries
  HIRS9202380
Transfer capillaries
Transfer capillaries

80 mm, 2,35 mm, 1,55

 
SFr. 120.00
   
1 * 1.000 ST
 

Clear glass, both end open.

 
  • Ext.Ø:
    2,35 mm
  • Ø int.:
    1,55
  • Length:
    80 mm
  • Seq:
    30
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.