Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 200-300 ml, 19/26
Katalognr. 215-1245
Anbieter: HIRSCHMANN
Direkt zu
Product Details & Documents
Oxygen bottles according to Winkler, with white inscription label.
- The adjusted capacity is individually marked on the bottle
- A unique serial number is engraved on the bottle and stopper
Technische Daten
- Capacity:200-300 ml
- Description:Oxygen bottles acc. to Winkler
- NS stopper:19/26