Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 100-150 ml, 14/23
Catalog # 612-2794
Supplier:   HIRSCHMANN
Dissolved oxygen bottles
Catalog # 612-2794
Supplier:  HIRSCHMANN
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 100-150 ml, 14/23
Price
42.40 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Oxygen bottles according to Winkler, with white inscription label.
  • The adjusted capacity is individually marked on the bottle
  • A unique serial number is engraved on the bottle and stopper
Specifications
  • Capacity:
    100-150 ml
  • Description:
    Oxygen bottles acc. to Winkler
  • NS stopper:
    14/23