Order Entry
Switzerland
Orders LinkContactUsLinkComponent
ICP multi-element standards

ICP multi-element standards

Multi-element calibration standard-4, 100 ml, (12 elements in H20/trace HF) B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr, 10 mg/L, 1% HNO₃ + (CHOH)₂(COOH)₂, 1
Catalog # HEWL8500-6942
ICP multi-element standards
Catalog # HEWL8500-6942
CAS Number:  
ICP multi-element standards
ICP multi-element standards

Multi-element calibration standard-4, 100 ml, (12 elements in H20/trace HF) B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr, 10 mg/L, 1% HNO₃ + (CHOH)₂(COOH)₂, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Documents

Specifications

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    1% HNO₃ + (CHOH)₂(COOH)₂
  • Pk:
    1
  • Description:
    Multi-element calibration standard-4, 100 ml, (12 elements in H<sub>2</sub>0/trace HF) B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.