ContactUsLinkComponent
STERISTOPPER, CELLULOSE, NUMBER 14, FOR GLASS WITH 13.0MM TO 14.5MM INNER DIAMETER 1 * 500 items
Catalog # 391-8914
Supplier:  HERENZ HEINZ
STERISTOPPER, CELLULOSE, NUMBER 14, FOR GLASS WITH 13.0MM TO 14.5MM INNER DIAMETER 1 * 500 items
Catalog # 391-8914
Supplier:  HERENZ HEINZ
Frequently Bought Together