Micro pipettes, disposable, with ring mark
1/2/3/4/5 µl, White, Five marks
Catalog # HECH40555003
Supplier: GLASWARENFABRIK KARL HECHT
Micro pipettes, disposable, with ring mark
Catalog # HECH40555003
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Disposable micro pipettes with ring mark.
- Calibrated to contain 'In'
- Colour-coded
: DIN ISO 7550
Specifications
- Colour code:White
- Type:Five marks
- Capacity:1/2/3/4/5 µl
- Seq:01