Electronic cell counters
Electronic cell counter, AC-15, 15
HECH40345015
The electronic memory devices specify blood counting.
- Power supply operation
- Rechargeable battery operation
- Software (download) with computer analysis
- Big TFT color display
- Storage of last counting on device
The microscope image can be observed permanently, the numbers can be typed in easily.
Colour: Blue
: Every device comes with an instruction manual.
- Model:AC-15
- Description:Electronic cell counter
- Seq:005
- Array:15
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



