Staining dishes
Staining dish without lid, 90×70×65 mm
Catalog # 630-2600
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Soda-lime glass.
- Removable cover
Staining dish for 76×26 mm slides or slides with width up to 45 mm.
Documents
HECH41205001_TDS_EN
Datasheet
Specifications
- Description:Staining dish without lid
- W×D×H:90×70×65 mm
Accessories
Product Information
- Catalog #W×D×HAvailabilityPrice
- Lid for staining dish
Product Information
- Catalog #W×D×HAvailabilityPrice
- Stainless steel rack for staining jars, for 10 slidesStainless steel rack for staining jars, for 10 slides631-0348- mmLog in for availability1 * 1 ST - SFr. 25.10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



