Orders LinkContactUsLinkComponent
Dissecting forceps
Dissecting forceps
Dissecting forceps, stainless steel, tip width 0,05 mm, tip thickness 0,01 mm, Straight, 110 mm
Catalog # HARV60-3854
Supplier:   HARVARD APPARATUS
Dissecting forceps
Catalog # HARV60-3854
Supplier:  HARVARD APPARATUS
Dissecting forceps
Dissecting forceps, stainless steel, tip width 0,05 mm, tip thickness 0,01 mm, Straight, 110 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Micro dissecting forceps with tower tip. Made from stainless steel or Inox steel.
Specifications
  • Type:
    Straight
  • Description:
    Dissecting forceps, stainless steel, tip width 0,05 mm, tip thickness 0,01 mm
  • Length:
    110 mm