Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Dissecting needles

Straight, 50 mm, 0,6 mm
  631-7174
Dissecting needles
  631-7174
Dissecting needles
Dissecting needles

Straight, 50 mm, 0,6 mm

 
SFr. 54.70
   
1 * 10 ST
 

Stainless steel 18/10, anti-magnetic.

 
  • Ø:
    0,6 mm
  • Form:
    Straight
  • Length:
    50 mm
  • Seq:
    03