Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dissecting needles

Dissecting needles

Lancet, 50 mm, 0,6 mm
Catalog # 631-7173
Supplier:   HAMMACHER KARL
Dissecting needles
Catalog # 631-7173
Supplier:  HAMMACHER KARL
Dissecting needles
Dissecting needles

Lancet, 50 mm, 0,6 mm

Price
56.60 SFr.
Log in for availability
1 * 10 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic.

Specifications
  • Ø:
    0,6 mm
  • Form:
    Lancet
  • Length:
    50 mm