Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Spatulas, micro

130 mm, 5 mm
Catalog # 231-1366
Supplier:   HAMMACHER KARL
Spatulas, micro
Catalog # 231-1366
Supplier:  HAMMACHER KARL
Spatulas, micro
Spatulas, micro

130 mm, 5 mm

Price
SFr. 24.70
Log in for availability
1 * 1 ST
Product Details & Documents

Single-ended micro spatulas made from WIRONIT steel.

Specifications
  • Length:
    130 mm
  • Seq:
    05
  • Blade width:
    5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.