Orders LinkContactUsLinkComponent
Dissecting needles
Dissecting needles
Lancet
Catalog # 233-0080
Supplier:   HAMMACHER KARL
Dissecting needles
Catalog # 233-0080
Supplier:  HAMMACHER KARL
Dissecting needles
Dissecting needles
Lancet
Price
SFr. 20.10
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel needle, with plastic handle.
Specifications
  • Form:
    Lancet