Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Section lifters

Section lifters

150 mm, 15 mm
Catalog # HAMMHSN444-15
Supplier:   HAMMACHER KARL
Section lifters
Catalog # HAMMHSN444-15
Supplier:  HAMMACHER KARL
Section lifters
Section lifters

150 mm, 15 mm

Price
SFr. 17.20
Log in for availability
1 * 1 ST
Product Details & Documents

Section lifters with flexible blade and wooden handle.

Specifications
  • Length:
    150 mm
  • Seq:
    06
  • Blade width:
    15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.