Orders LinkContactUsLinkComponent
Microscopic forceps
Microscopic forceps
Microscopic forceps, Straight, 105 mm
Catalog # HAMMHSC550-10
Supplier:   HAMMACHER KARL
Microscopic forceps
Catalog # HAMMHSC550-10
Supplier:  HAMMACHER KARL
Microscopic forceps
Microscopic forceps
Microscopic forceps, Straight, 105 mm
Price
64.20 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Straight
  • Description:
    Microscopic forceps
  • Length:
    105 mm
  • Seq:
    01