BUFFER PH 9,21 DOSING BOTTLE 1 * 500 mL
Catalog # HAMI238219
Supplier: HAMILTON BONADUZ
Frequently Bought Together
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Syringes, 3-piece
Supplier: TERUMO
Acrodisc® One PSF Syringe Filters, wwPTFE Membrane
Supplier: Cytiva (Formerly Pall Lab)
BUFFER PH 10 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Funnels with Short Stem
Supplier: VWR Collection
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Supplier: HAMILTON BONADUZ
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Single use syringes, 2-piece, Injekt® Solo
Supplier: B. Braun
Samplers for liquids, TeleScoop
Supplier: BURKLE
VWR® Bulb Pipettes, Two Marks, Class AS
Supplier: VWR Collection
VWR®, Twist-Seal Bags
Supplier: VWR Collection
Minigrip® Bags, Zip Seal, Resistant, Euro-Hole
Supplier: Flexico
Beakers, low or tall form
Supplier: Kavalier
Petri dishes
Supplier: Greiner Bio-One
Silver nitrate 0.1 M AgNO3 (0.1N) in aqueous solution ≥99% Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Buffer Solutions
Supplier: Thermo Fisher Scientific
Buffer solutions
Supplier: REAGECON