Colour standards
Standard colour solution, 500 Pt-Co units, Bottle, 1× 1 litre, 1 L
Supplier: Hach
Jump to:
Product Details & Documents
For accuracy checks in colour determinations.
- Simple Accuracy Checks
- Minimize waste with Voluette ampoules
- Reliable results
The voluette format has the following advantages: Open a fresh standard amopule for each accuracy check and each ampule contains enough standard for one set of standard additions. Sealed ampoules prevent accidental contamination or evaporation of standard solutions.
Documents
Specifications
Specifications
- Pack type:Bottle
- Pk:1 L
- Description:Standard colour solution, 500 Pt-Co units
- Pk Info:1× 1 litre