Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Hydrometers

Hydrometers

1,400 - 1,500 g/ml, 0,001 g/ml
Catalog # GERI15/1.4-1.5
Supplier:   GERING
Hydrometers
Catalog # GERI15/1.4-1.5
Supplier:  GERING
Hydrometers
Hydrometers

1,400 - 1,500 g/ml, 0,001 g/ml

Price
SFr. 60.50
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Seq:
    09
  • Division:
    0,001 g/ml
  • Range:
    1,400 - 1,500 g/ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.