Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Colour Reference Solutions, Reag.Ph.Eur.

Colour Reference Solutions, Reag.Ph.Eur.

Colour reference solution R (red) for testing the colour intensity acc. to Ph. Eur. R1-R7 Reag. Ph. Eur., Glass bottle, 125 mL
Catalog # REGEEP707
Supplier:   REAGECON
Colour Reference Solutions, Reag.Ph.Eur.
Catalog # REGEEP707
Supplier:  REAGECON
CAS Number:  
Colour Reference Solutions, Reag.Ph.Eur.
Colour Reference Solutions, Reag.Ph.Eur.

Colour reference solution R (red) for testing the colour intensity acc. to Ph. Eur. R1-R7 Reag. Ph. Eur., Glass bottle, 125 mL

CAS Number:  
Price
226.00 SFr.
Log in for availability
1 * 125 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Description:
    Colour reference solution R (red) for testing the colour intensity acc. to Ph. Eur. R1-R7 Reag. Ph. Eur.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.