ContactUsLinkComponent
Coelenterata and Porifera. - 10 Microscope Slides - 1. Sycon, a small marine sponge of the sycon type, l.s. and t.s. on one slide 2. Spongilla, fresh-water sponge, t.s. 3. Euspongia, commercial sponge, t.s. 4. Sponge spicules of different kinds, mix 1 * 1 items
Catalog # LIED74600N
Supplier:  LIEDER JOHANNES
Coelenterata and Porifera. - 10 Microscope Slides - 1. Sycon, a small marine sponge of the sycon type, l.s. and t.s. on one slide 2. Spongilla, fresh-water sponge, t.s. 3. Euspongia, commercial sponge, t.s. 4. Sponge spicules of different kinds, mix 1 * 1 items
Catalog # LIED74600N
Supplier:  LIEDER JOHANNES
Frequently Bought Together