Orders LinkContactUsLinkComponent
Accessories for Axio Vert. A1
Accessories for Axio Vert. A1
Mounting frame for microplates, 60, 72 and 120 wells
Catalog # 630-0413
Supplier:   ZEISS
Accessories for Axio Vert. A1
Catalog # 630-0413
Supplier:  ZEISS
Accessories for Axio Vert. A1
Accessories for Axio Vert. A1
Mounting frame for microplates, 60, 72 and 120 wells
Price
361.95 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Mounting frame for microplates, 60, 72 and 120 wells
  • Seq:
    04