Orders LinkContactUsLinkComponent
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
SACIQ-7,0 cable for all IQ sensors with IP68 waterproof connection to the IQ Sensor Net system, 7 m
Catalog # 708-0048
Supplier:   WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Catalog # 708-0048
Supplier:  WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
SACIQ-7,0 cable for all IQ sensors with IP68 waterproof connection to the IQ Sensor Net system, 7 m
Price
280.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    SACIQ-7,0 cable for all IQ sensors with IP68 waterproof connection to the IQ Sensor Net system, 7 m
  • Seq:
    02