ContactUsLinkComponent
Ua 55,universal extension assembly (incl. handle and set of seals) for mounting of sensors of series 650/690/70x und 70x iq, length without sensor: 317 mm 1 * 1 items
  662-0507
Ua 55,universal extension assembly (incl. handle and set of seals) for mounting of sensors of series 650/690/70x und 70x iq, length without sensor: 317 mm 1 * 1 items
  662-0507
Frequently Bought Together