Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sample vials in Lab File®, WHEATON®

Sample vials in Lab File®, WHEATON®

8,00 ml, 17×60 mm, Clear, No, 15-425
Catalog # 218-2223
Supplier:   DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Catalog # 218-2223
Supplier:  DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Sample vials in Lab File®, WHEATON®

8,00 ml, 17×60 mm, Clear, No, 15-425

Price
SFr. 255.00
Log in for availability
1 * 200 ST
Product Details & Documents

For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.

Ordering information: Without cap.

Specifications
  • Cap included:
    No
  • Cap Size:
    15-425
  • Ø×H:
    17×60 mm
  • Capacity:
    8,00 ml
  • Colour:
    Clear
  • Seq:
    07