Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Sample vials in Lab File®, WHEATON®

2,00 ml, 12×35 mm, Clear, No, 8-425
  218-2221
Sample vials in Lab File®, WHEATON®
  218-2221
Sample vials in Lab File®, WHEATON®
Sample vials in Lab File®, WHEATON®

2,00 ml, 12×35 mm, Clear, No, 8-425

 
SFr. 206.00
   
1 * 200 ST
 

For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.

 : Without cap.

 
  • Cap included:
    No
  • Cap Size:
    8-425
  • Ø×H:
    12×35 mm
  • Capacity:
    2,00 ml
  • Colour:
    Clear
  • Seq:
    05