Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Intelsius® Pathopouch

Intelsius® Pathopouch

95 kPa pouch for pathology, A3, 359×261 mm
Catalog # DGINBP005
New Product
Supplier:   DGP INTELSIUS
Intelsius® Pathopouch
Catalog # DGINBP005
Supplier:  DGP INTELSIUS
New Product
Intelsius® Pathopouch
Intelsius® Pathopouch

95 kPa pouch for pathology, A3, 359×261 mm

Price
Inquire For Price
Log in for availability
1 * 50 ST
Product Details & Documents

The Intelsius® Pathopouch range are 95 kPa flexible pressure vessels designed to meet with transport regulations.

  • 95 kPa plastic secondary packaging

  • Outer pouch for paperwork
  • Leak-proof

They are leak-proof and suitable to hold a wide range of primary containers, such as blood vials, specimen containers, swabs, blood collection bags and medical devices.

Certifications: ISTA Certified Class 6.2 Category A & B

Specifications
  • Description:
    95 kPa pouch for pathology
  • L×W:
    359×261 mm
  • Seq:
    2
  • Size:
    A3
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.