EasyPlus™ Titration Electrodes
EG11-BNC sensor pH aqueous, ±2000 mV/0 - 14 pH, Easy Pro, Easy EPM, Easy pH titrators
Catalog # METT30043103
New Product
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus™ titrators only.
Electrode only.
Specifications
- For:Easy Pro, Easy EPM, Easy pH titrators
- Description:EG11-BNC sensor pH aqueous
- Seq:1
- Range:±2000 mV/0 - 14 pH