Combination Blocks
CCT-W09-COMBO block, Ø 11,1/17,1/29 mm, 1,5/15/50 ml conical centrifuge, 4/3/2, 39,1/44,5/47,6 mm
Catalog # VELPA00000512
New Product
Supplier: VELP SCIENTIFIC
Jump to:
Product Details & Documents
Specifications
- No. of Holes:4/3/2
- Description:CCT-W09-COMBO block
- Seq:2
- Depth:39,1/44,5/47,6 mm
- Well size:Ø 11,1/17,1/29 mm
- For:1,5/15/50 ml conical centrifuge